Product Details

SNP ID
rs141566500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66908435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACGCTGTCTGCTGGTTGATCC[A/G]GGTCCTTCTTCCTTGACATGGTCCA
Phenotype
MIM: 603118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH16 PubMed Links

Gene Details

Gene
CDH16
Gene Name
cadherin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204744.1 2532 Missense Mutation NP_001191673.1
NM_001204745.1 2532 Missense Mutation NP_001191674.1
NM_001204746.1 2532 Missense Mutation NP_001191675.1
NM_004062.3 2532 Missense Mutation NP_004053.1
XM_005255770.2 2532 Missense Mutation XP_005255827.1
XM_011522807.1 2532 Missense Mutation XP_011521109.1

View Full Product Details