Product Details

SNP ID
rs142058878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:631325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTCGGGGGCTGGCTTGCTGCC[A/G]GGGCTGGGGAGCCACCCGGGCGTGT
Phenotype
MIM: 608021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100287175 PubMed Links

Gene Details

Gene
LOC100287175
Gene Name
uncharacterized LOC100287175
There are no transcripts associated with this gene.

Gene
METTL26
Gene Name
methyltransferase like 26
There are no transcripts associated with this gene.

Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
WFIKKN1
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053284.2 314 Silent Mutation CCA,CCG P24P NP_444514.1

View Full Product Details