Product Details

SNP ID
rs142158761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28956165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACTGCAGTGAGGTGAACAAGC[A/G]CCTCCAGGATCTCCGTTCCTGTCTG
Phenotype
MIM: 614525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4517 PubMed Links

Gene Details

Gene
MIR4517
Gene Name
microRNA 4517
There are no transcripts associated with this gene.

Gene
NFATC2IP
Gene Name
nuclear factor of activated T-cells 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032815.3 689 Missense Mutation CAC,CGC H225R NP_116204.3

View Full Product Details