Product Details

SNP ID
rs143109128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58516647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCCCTAGAACTCTACCACCAG[C/G]TGGTGGCCCTTGTGATGGCCTATAG
Phenotype
MIM: 604917 MIM: 614463 MIM: 616424
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNOT1 PubMed Links

Gene Details

Gene
CNOT1
Gene Name
CCR4-NOT transcription complex subunit 1
There are no transcripts associated with this gene.

Gene
NDRG4
Gene Name
NDRG family member 4
There are no transcripts associated with this gene.

Gene
SETD6
Gene Name
SET domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160305.2 633 Silent Mutation CTG,GTG L216V NP_001153777.1
NM_024860.3 633 Silent Mutation CTG,GTG L192V NP_079136.2
XM_011523336.1 633 Missense Mutation CTG,GTG L216V XP_011521638.1

View Full Product Details