Product Details

SNP ID
rs143156767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66923718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCCGGGCAACCAGCGGCCCCC[A/G]TCCTGGAGAACACACAACACACATC
Phenotype
MIM: 603118 MIM: 614778 MIM: 179503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH16 PubMed Links

Gene Details

Gene
CDH16
Gene Name
cadherin 16
There are no transcripts associated with this gene.

Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
There are no transcripts associated with this gene.

Gene
RRAD
Gene Name
RRAD, Ras related glycolysis inhibitor and calcium channel regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128850.1 590 Silent Mutation GAC,GAT D149D NP_001122322.1
NM_004165.2 590 Silent Mutation GAC,GAT D149D NP_004156.1

View Full Product Details