Product Details
- SNP ID
-
rs145141523
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75229918 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGCTGCCGGTCCGAGGGCCCCA[A/G]GCCGCCTGTTCGCCCCGGGGCCAGG
- Phenotype
-
MIM: 602941
MIM: 118890
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCAR1
PubMed Links
Gene Details
- Gene
- BCAR1
- Gene Name
- BCAR1, Cas family scaffolding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170714.1 |
2780 |
Silent Mutation |
CTG,TTG |
L782L |
NP_001164185.1 |
NM_001170715.1 |
2780 |
Silent Mutation |
CTG,TTG |
L754L |
NP_001164186.1 |
NM_001170716.1 |
2780 |
Silent Mutation |
CTG,TTG |
L754L |
NP_001164187.1 |
NM_001170717.1 |
2780 |
Silent Mutation |
CTG,TTG |
L754L |
NP_001164188.1 |
NM_001170718.1 |
2780 |
Silent Mutation |
CTG,TTG |
L736L |
NP_001164189.1 |
NM_001170719.1 |
2780 |
Silent Mutation |
CTG,TTG |
L734L |
NP_001164190.1 |
NM_001170720.1 |
2780 |
Silent Mutation |
CTG,TTG |
L588L |
NP_001164191.1 |
NM_001170721.1 |
2780 |
Silent Mutation |
CTG,TTG |
L526L |
NP_001164192.1 |
NM_014567.3 |
2780 |
Silent Mutation |
CTG,TTG |
L736L |
NP_055382.2 |
XM_005256258.4 |
2780 |
Silent Mutation |
CTG,TTG |
L762L |
XP_005256315.1 |
XM_005256259.4 |
2780 |
Silent Mutation |
CTG,TTG |
L736L |
XP_005256316.1 |
XM_005256260.4 |
2780 |
Silent Mutation |
CTG,TTG |
L707L |
XP_005256317.1 |
XM_011523475.2 |
2780 |
Silent Mutation |
CTG,TTG |
L707L |
XP_011521777.1 |
XM_017023897.1 |
2780 |
Silent Mutation |
CTG,TTG |
L782L |
XP_016879386.1 |
XM_017023898.1 |
2780 |
Silent Mutation |
CTG,TTG |
L752L |
XP_016879387.1 |
XM_017023899.1 |
2780 |
Silent Mutation |
CTG,TTG |
L689L |
XP_016879388.1 |
- Gene
- CTRB1
- Gene Name
- chymotrypsinogen B1
There are no transcripts associated with this gene.
- Gene
- LOC100506281
- Gene Name
- uncharacterized LOC100506281
There are no transcripts associated with this gene.
View Full Product Details