Product Details

SNP ID
rs145758365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30604508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCGCTCGCCCGAGTGCACGCGC[C/T]GGTGGCTGGCCAGCAGTGAGGGGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF689 PubMed Links

Gene Details

Gene
ZNF689
Gene Name
zinc finger protein 689
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138447.2 1604 Missense Mutation CCG,CTG P420L NP_612456.1

View Full Product Details