Product Details

SNP ID
rs146436156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:676706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGCTCCAGCAGTTTCAAGCGGG[C/T]CATTGCTAACGGACAGCGGGCACTG
Phenotype
MIM: 603264 MIM: 613889 MIM: 607207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD8 PubMed Links

Gene Details

Gene
JMJD8
Gene Name
jumonji domain containing 8
There are no transcripts associated with this gene.

Gene
LOC105371184
Gene Name
uncharacterized LOC105371184
There are no transcripts associated with this gene.

Gene
RHBDL1
Gene Name
rhomboid like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278720.1 461 Missense Mutation GCC,GTC A79V NP_001265649.1
NM_001278721.1 461 Intron NP_001265650.1
NM_001318733.1 461 Missense Mutation GCC,GTC A144V NP_001305662.1
XM_017023849.1 461 Missense Mutation GCC,GTC A144V XP_016879338.1
XM_017023850.1 461 Missense Mutation GCC,GTC A144V XP_016879339.1
Gene
RHOT2
Gene Name
ras homolog family member T2
There are no transcripts associated with this gene.

Gene
STUB1
Gene Name
STIP1 homology and U-box containing protein 1
There are no transcripts associated with this gene.

Gene
WDR24
Gene Name
WD repeat domain 24
There are no transcripts associated with this gene.

Gene
WDR90
Gene Name
WD repeat domain 90
There are no transcripts associated with this gene.

View Full Product Details