Product Details

SNP ID
rs149489369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56935273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTTAATTTATTCTGGGAAGCT[A/G]TTGTTGGATCACCAATGTCTCAGGG
Phenotype
MIM: 608070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HERPUD1 PubMed Links

Gene Details

Gene
HERPUD1
Gene Name
homocysteine inducible ER protein with ubiquitin like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010989.2 341 Silent Mutation CTA,CTG L62L NP_001010989.1
NM_001272103.1 341 Silent Mutation CTA,CTG L62L NP_001259032.1
NM_014685.3 341 Silent Mutation CTA,CTG L62L NP_055500.1

View Full Product Details