Product Details

SNP ID
rs149577827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67876050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTGATAGTACCTCAGCAAACA[A/G]GACTGGTCTTCGGACCATGCCACCC
Phenotype
MIM: 606030 MIM: 605813
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDC4 PubMed Links

Gene Details

Gene
EDC4
Gene Name
enhancer of mRNA decapping 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014329.4 427 Missense Mutation AAG,AGG K63R NP_055144.3
Gene
NRN1L
Gene Name
neuritin 1 like
There are no transcripts associated with this gene.

Gene
NUTF2
Gene Name
nuclear transport factor 2
There are no transcripts associated with this gene.

View Full Product Details