Product Details

SNP ID
rs151237402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31717334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGACATTCTTTCTCTAGGACTG[A/G]GAATCATTGATTTGAACTATAGGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLUHP3 PubMed Links

Gene Details

Gene
CLUHP3
Gene Name
clustered mitochondria homolog pseudogene 3
There are no transcripts associated with this gene.

Gene
ZNF720
Gene Name
zinc finger protein 720
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130913.1 Intron NP_001124385.1

View Full Product Details