Product Details

SNP ID
rs138536120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2605324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGCAGAGAATAAGAGGAGGAG[C/T]TGTATAGAATATTACATAAAGCAGC
Phenotype
MIM: 601545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAFAH1B1 PubMed Links

Gene Details

Gene
PAFAH1B1
Gene Name
platelet activating factor acetylhydrolase 1b regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000430.3 Intron NP_000421.1
XM_011523901.1 Intron XP_011522203.1
XM_011523902.2 Intron XP_011522204.1
XM_011523903.2 Intron XP_011522205.1
XM_017024699.1 Intron XP_016880188.1
XM_017024700.1 Intron XP_016880189.1
XM_017024701.1 Intron XP_016880190.1
XM_017024702.1 Intron XP_016880191.1
XM_017024703.1 Intron XP_016880192.1

View Full Product Details