Product Details

SNP ID
rs138621912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82025453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTCCCCAGCAACAGAACCCTG[G/T]GGAGACTGGACCTGGCTGGGAACAA
Phenotype
MIM: 606236 MIM: 602050 MIM: 615128
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
There are no transcripts associated with this gene.

Gene
LRRC45
Gene Name
leucine rich repeat containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144999.3 949 Missense Mutation GGG,TGG G203W NP_659436.1
Gene
RAC3
Gene Name
ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3)
There are no transcripts associated with this gene.

Gene
STRA13
Gene Name
stimulated by retinoic acid 13
There are no transcripts associated with this gene.

View Full Product Details