Product Details

SNP ID
rs138644253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75266360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCTTATACAAAATGTATAAAA[A/G]GCAGTTTCTGGTGTGACTTGTGCTC
Phenotype
MIM: 606006 MIM: 612072 MIM: 611974 MIM: 606521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGA3 PubMed Links

Gene Details

Gene
GGA3
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
There are no transcripts associated with this gene.

Gene
LOC100287042
Gene Name
uncharacterized LOC100287042
There are no transcripts associated with this gene.

Gene
MIF4GD
Gene Name
MIF4G domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242498.1 1879 UTR 3 NP_001229427.1
NM_001242500.1 1879 UTR 3 NP_001229429.1
NM_001242501.1 1879 UTR 3 NP_001229430.1
NM_020679.3 1879 UTR 3 NP_065730.2
XM_005257530.1 1879 UTR 3 XP_005257587.1
XM_005257532.4 1879 UTR 3 XP_005257589.1
XM_005257533.1 1879 UTR 3 XP_005257590.1
XM_011525051.2 1879 UTR 3 XP_011523353.1
XM_011525052.2 1879 UTR 3 XP_011523354.1
XM_011525053.2 1879 UTR 3 XP_011523355.1
XM_011525054.2 1879 UTR 3 XP_011523356.1
XM_011525055.2 1879 UTR 3 XP_011523357.1
XM_011525057.2 1879 UTR 3 XP_011523359.1
XM_017024888.1 1879 UTR 3 XP_016880377.1
XM_017024889.1 1879 UTR 3 XP_016880378.1
XM_017024890.1 1879 UTR 3 XP_016880379.1
XM_017024891.1 1879 UTR 3 XP_016880380.1
XM_017024892.1 1879 Intron XP_016880381.1
XM_017024893.1 1879 UTR 3 XP_016880382.1
XM_017024894.1 1879 UTR 3 XP_016880383.1
XM_017024895.1 1879 Intron XP_016880384.1
Gene
MRPS7
Gene Name
mitochondrial ribosomal protein S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015971.3 1879 UTR 3 NP_057055.2
Gene
SLC25A19
Gene Name
solute carrier family 25 member 19
There are no transcripts associated with this gene.

View Full Product Details