Product Details

SNP ID
rs139455952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76275904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGAGGGATGGCGGCCTGTGCAC[A/G]TGGGGCCTGGGCTGCTGGGACTTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QRICH2 PubMed Links

Gene Details

Gene
QRICH2
Gene Name
glutamine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032134.2 6089 Silent Mutation NP_115510.1
XM_005257728.3 6089 Silent Mutation XP_005257785.1
XM_006722136.3 6089 Silent Mutation XP_006722199.1
XM_011525344.2 6089 Silent Mutation XP_011523646.1
XM_017025206.1 6089 Missense Mutation XP_016880695.1
XM_017025207.1 6089 Silent Mutation XP_016880696.1
XM_017025208.1 6089 Silent Mutation XP_016880697.1
Gene
UBALD2
Gene Name
UBA like domain containing 2
There are no transcripts associated with this gene.

View Full Product Details