Product Details

SNP ID
rs139863746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76274237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGACGGCCCTCGGAGGAAGGTC[C/T]ATCTTTCTGTTGACGAGAAGAAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
QRICH2 PubMed Links

Gene Details

Gene
QRICH2
Gene Name
glutamine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005257728.3 7498 Missense Mutation AGA,GGA R1836G XP_005257785.1
XM_006722136.3 7498 Missense Mutation AGA,GGA R590G XP_006722199.1
XM_011525344.2 7498 Missense Mutation AGA,GGA R1596G XP_011523646.1
XM_017025206.1 7498 UTR 3 XP_016880695.1
XM_017025207.1 7498 Missense Mutation AGA,GGA R1816G XP_016880696.1
XM_017025208.1 7498 Missense Mutation AGA,GGA R514G XP_016880697.1
Gene
UBALD2
Gene Name
UBA like domain containing 2
There are no transcripts associated with this gene.

View Full Product Details