Product Details

SNP ID
rs140293241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39651990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACATGGCAGGGAGGGAGTGCCA[A/G]AGTTCCTTTTGTCTTTAGGGTTCCT
Phenotype
MIM: 607048
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STARD3 PubMed Links

Gene Details

Gene
STARD3
Gene Name
StAR related lipid transfer domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165937.1 Intron NP_001159409.1
NM_001165938.1 Intron NP_001159410.1
NM_006804.3 Intron NP_006795.3
XM_006721646.2 Intron XP_006721709.1
XM_011524210.1 Intron XP_011522512.1
XM_011524211.2 Intron XP_011522513.1
XM_011524213.1 Intron XP_011522515.1
XM_017024041.1 Intron XP_016879530.1

View Full Product Details