Product Details

SNP ID
rs140416514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45149255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACTAGCAACTGTACAGGTGCTG[C/T]TGCTGTCCAGGAAGAGCTGAACCCT
Phenotype
MIM: 607328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACBD4 PubMed Links

Gene Details

Gene
ACBD4
Gene Name
acyl-CoA binding domain containing 4
There are no transcripts associated with this gene.

Gene
HEXIM1
Gene Name
hexamethylene bisacetamide inducible 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006460.2 1939 Missense Mutation GCT,GTT A22V NP_006451.1

View Full Product Details