Product Details

SNP ID
rs141915492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17496040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGCGTGTAGGCGTCCTCGAAGC[G/T]GCCGGTGAGGAAGCGCGACACGATG
Phenotype
MIM: 609878 MIM: 602391 MIM: 605550
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MED9 PubMed Links

Gene Details

Gene
MED9
Gene Name
mediator complex subunit 9
There are no transcripts associated with this gene.

Gene
PEMT
Gene Name
phosphatidylethanolamine N-methyltransferase
There are no transcripts associated with this gene.

Gene
RASD1
Gene Name
ras related dexamethasone induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199989.1 356 Missense Mutation NP_001186918.1
NM_016084.4 356 Missense Mutation NP_057168.1

View Full Product Details