Product Details

SNP ID
rs143018406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42815975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCCAGCTGCGTCTGGGCATAAC[A/G]CATCTGGTTTTCAACACTCTTCAGC
Phenotype
MIM: 604378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BECN1 PubMed Links

Gene Details

Gene
BECN1
Gene Name
beclin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313998.1 765 Missense Mutation NP_001300927.1
NM_001313999.1 765 Missense Mutation NP_001300928.1
NM_001314000.1 765 Missense Mutation NP_001300929.1
NM_003766.4 765 Missense Mutation NP_003757.1
XM_005257759.2 765 Missense Mutation XP_005257816.1
XM_005257760.3 765 Missense Mutation XP_005257817.1
XM_011525421.2 765 Missense Mutation XP_011523723.1
XM_017025262.1 765 Missense Mutation XP_016880751.1
XM_017025263.1 765 Missense Mutation XP_016880752.1
XM_017025264.1 765 Missense Mutation XP_016880753.1
Gene
CNTD1
Gene Name
cyclin N-terminal domain containing 1
There are no transcripts associated with this gene.

Gene
MIR6781
Gene Name
microRNA 6781
There are no transcripts associated with this gene.

View Full Product Details