Product Details

SNP ID
rs145094200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1423620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTTCCCACTGACCACTCACAT[C/T]AATCTTCGTAACCTTTACCAGCTCA
Phenotype
MIM: 164762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005206.4 958 UTR 3 NP_005197.3
NM_016823.3 958 Missense Mutation AAT,GAT N270D NP_058431.2

View Full Product Details