Product Details

SNP ID
rs145177186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17495193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGGCCTTCTCGCGGATGTACA[C/T]GAGGTCGCTGTGTACGCTGGGCCGG
Phenotype
MIM: 609878 MIM: 605550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED9 PubMed Links

Gene Details

Gene
MED9
Gene Name
mediator complex subunit 9
There are no transcripts associated with this gene.

Gene
RASD1
Gene Name
ras related dexamethasone induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199989.1 918 UTR 3 NP_001186918.1
NM_016084.4 918 Missense Mutation ATG,GTG M260V NP_057168.1

View Full Product Details