Product Details

SNP ID
rs146952597
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19773678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGAGGGATGGATGCTATCCT[A/T]CTCTGGACTGAAGCAGTTACAGGAT
Phenotype
MIM: 608650
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ULK2 PubMed Links

Gene Details

Gene
ULK2
Gene Name
unc-51 like autophagy activating kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142610.1 6291 Intron NP_001136082.1
NM_014683.3 6291 UTR 3 NP_055498.3
XM_011524087.2 6291 Intron XP_011522389.1
XM_017025424.1 6291 Intron XP_016880913.1
XM_017025425.1 6291 Intron XP_016880914.1
XM_017025426.1 6291 Intron XP_016880915.1
XM_017025427.1 6291 Intron XP_016880916.1
XM_017025428.1 6291 Intron XP_016880917.1

View Full Product Details