Product Details
- SNP ID
-
rs148422968
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75267783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGATGTTGCAGATAAAGGTGACA[A/T]AGCAGACCCAGCCCTGCAGGGAGCG
- Phenotype
-
MIM: 606006
MIM: 612072
MIM: 611974
MIM: 606521
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GGA3
PubMed Links
Gene Details
- Gene
- GGA3
- Gene Name
- golgi associated, gamma adaptin ear containing, ARF binding protein 3
There are no transcripts associated with this gene.
- Gene
- LOC100287042
- Gene Name
- uncharacterized LOC100287042
There are no transcripts associated with this gene.
- Gene
- MIF4GD
- Gene Name
- MIF4G domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242498.1 |
571 |
Missense Mutation |
TAT,TTT |
Y145F |
NP_001229427.1 |
NM_001242500.1 |
571 |
Missense Mutation |
TAT,TTT |
Y138F |
NP_001229429.1 |
NM_001242501.1 |
571 |
Missense Mutation |
TAT,TTT |
Y104F |
NP_001229430.1 |
NM_020679.3 |
571 |
Missense Mutation |
TAT,TTT |
Y138F |
NP_065730.2 |
XM_005257530.1 |
571 |
Missense Mutation |
TAT,TTT |
Y145F |
XP_005257587.1 |
XM_005257532.4 |
571 |
Missense Mutation |
TAT,TTT |
Y104F |
XP_005257589.1 |
XM_005257533.1 |
571 |
Missense Mutation |
TAT,TTT |
Y104F |
XP_005257590.1 |
XM_011525051.2 |
571 |
Missense Mutation |
TAT,TTT |
Y213F |
XP_011523353.1 |
XM_011525052.2 |
571 |
Missense Mutation |
TAT,TTT |
Y206F |
XP_011523354.1 |
XM_011525053.2 |
571 |
Missense Mutation |
TAT,TTT |
Y172F |
XP_011523355.1 |
XM_011525054.2 |
571 |
Intron |
|
|
XP_011523356.1 |
XM_011525055.2 |
571 |
Intron |
|
|
XP_011523357.1 |
XM_011525057.2 |
571 |
Intron |
|
|
XP_011523359.1 |
XM_017024888.1 |
571 |
Missense Mutation |
TAT,TTT |
Y213F |
XP_016880377.1 |
XM_017024889.1 |
571 |
Missense Mutation |
TAT,TTT |
Y145F |
XP_016880378.1 |
XM_017024890.1 |
571 |
Missense Mutation |
TAT,TTT |
Y145F |
XP_016880379.1 |
XM_017024891.1 |
571 |
Missense Mutation |
TAT,TTT |
Y145F |
XP_016880380.1 |
XM_017024892.1 |
571 |
Intron |
|
|
XP_016880381.1 |
XM_017024893.1 |
571 |
Missense Mutation |
TAT,TTT |
Y104F |
XP_016880382.1 |
XM_017024894.1 |
571 |
Missense Mutation |
TAT,TTT |
Y104F |
XP_016880383.1 |
XM_017024895.1 |
571 |
Intron |
|
|
XP_016880384.1 |
- Gene
- MRPS7
- Gene Name
- mitochondrial ribosomal protein S7
There are no transcripts associated with this gene.
- Gene
- SLC25A19
- Gene Name
- solute carrier family 25 member 19
There are no transcripts associated with this gene.
View Full Product Details