Product Details

SNP ID
rs149295671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50185590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCCACATCGATGATGGGCAGG[C/T]GGGAGGTCTTGGTGGTTTTGTATTC
Phenotype
MIM: 120150 MIM: 600119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL1A1 PubMed Links

Gene Details

Gene
COL1A1
Gene Name
collagen type I alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000088.3 4244 Missense Mutation CAC,CGC H1436R NP_000079.2
XM_005257058.3 4244 Missense Mutation CAC,CGC H1346R XP_005257115.2
XM_005257059.4 4244 Missense Mutation CAC,CGC H1130R XP_005257116.2
XM_011524341.1 4244 Missense Mutation CAC,CGC H1370R XP_011522643.1
Gene
SGCA
Gene Name
sarcoglycan alpha
There are no transcripts associated with this gene.

View Full Product Details