Product Details

SNP ID
rs150039501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37520634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACATACAGGAGGAAAAATCCAG[C/T]GAGTTTTCATCTGGCTGTGAGGACG
Phenotype
MIM: 606618 MIM: 607291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP14 PubMed Links

Gene Details

Gene
DUSP14
Gene Name
dual specificity phosphatase 14
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163544.2 3924 Silent Mutation NP_001157016.1
NM_001163545.2 3924 Silent Mutation NP_001157017.1
NM_001163546.2 3924 Silent Mutation NP_001157018.1
NM_001163547.2 3924 Silent Mutation NP_001157019.1
NM_007247.5 3924 Silent Mutation NP_009178.3
NM_080550.4 3924 Silent Mutation NP_542117.3
NM_198882.2 3924 Silent Mutation NP_942583.1
XM_005256980.4 3924 Silent Mutation XP_005257037.1
XM_017024076.1 3924 Silent Mutation XP_016879565.1
XM_017024077.1 3924 Silent Mutation XP_016879566.1
XM_017024078.1 3924 Silent Mutation XP_016879567.1
XM_017024079.1 3924 Silent Mutation XP_016879568.1
XM_017024080.1 3924 Silent Mutation XP_016879569.1
XM_017024081.1 3924 Silent Mutation XP_016879570.1
XM_017024082.1 3924 Silent Mutation XP_016879571.1
XM_017024083.1 3924 Silent Mutation XP_016879572.1
XM_017024084.1 3924 Silent Mutation XP_016879573.1
XM_017024085.1 3924 Silent Mutation XP_016879574.1
XM_017024086.1 3924 Silent Mutation XP_016879575.1
XM_017024087.1 3924 Silent Mutation XP_016879576.1
XM_017024088.1 3924 Silent Mutation XP_016879577.1
XM_017024089.1 3924 Silent Mutation XP_016879578.1
XM_017024090.1 3924 Silent Mutation XP_016879579.1
XM_017024091.1 3924 Silent Mutation XP_016879580.1
XM_017024092.1 3924 Silent Mutation XP_016879581.1
XM_017024093.1 3924 Silent Mutation XP_016879582.1
XM_017024094.1 3924 Silent Mutation XP_016879583.1
XM_017024095.1 3924 Silent Mutation XP_016879584.1
XM_017024096.1 3924 UTR 3 XP_016879585.1
XM_017024097.1 3924 Silent Mutation XP_016879586.1
XM_017024098.1 3924 Silent Mutation XP_016879587.1
XM_017024099.1 3924 Silent Mutation XP_016879588.1
XM_017024100.1 3924 Silent Mutation XP_016879589.1
XM_017024101.1 3924 Silent Mutation XP_016879590.1
XM_017024102.1 3924 Silent Mutation XP_016879591.1
XM_017024103.1 3924 Silent Mutation XP_016879592.1
XM_017024104.1 3924 Silent Mutation XP_016879593.1
XM_017024105.1 3924 Intron XP_016879594.1
XM_017024106.1 3924 Silent Mutation XP_016879595.1

View Full Product Details