Product Details

SNP ID
rs150602558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5002081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTGTGAGGACCTCTTCTCTC[A/G]CGTTAGTGAGAACCAGAGTGCTCAG
Phenotype
MIM: 603060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INCA1 PubMed Links

Gene Details

Gene
INCA1
Gene Name
inhibitor of CDK, cyclin A1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167985.1 734 Intron NP_001161457.1
NM_001167986.1 734 Intron NP_001161458.1
NM_001167987.1 734 Intron NP_001161459.1
NM_213726.2 734 Intron NP_998891.2
Gene
KIF1C
Gene Name
kinesin family member 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006612.5 734 Missense Mutation CAC,CGC H129R NP_006603.2
XM_005256424.2 734 Missense Mutation CAC,CGC H129R XP_005256481.1

View Full Product Details