Product Details

SNP ID
rs151181135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80470755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGAGAAGAGACGCACAAAACA[C/G]ATCATCGCCGCACAAGCAGGGGGCG
Phenotype
MIM: 602367
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NPTX1 PubMed Links

Gene Details

Gene
NPTX1
Gene Name
neuronal pentraxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002522.3 1515 UTR 3 NP_002513.2

View Full Product Details