Product Details

SNP ID
rs139987131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57545569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCTTTGGCAAGATGCATCAGAC[A/G]CACGGAGCGATGGTCTGGCTGAAGG
Phenotype
MIM: 612386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FECH PubMed Links

Gene Details

Gene
FECH
Gene Name
ferrochelatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000140.3 6550 UTR 3 NP_000131.2
NM_001012515.2 6550 UTR 3 NP_001012533.1
XM_011525881.1 6550 Intron XP_011524183.1
XM_011525882.2 6550 Intron XP_011524184.1
XM_017025614.1 6550 Intron XP_016881103.1

View Full Product Details