Product Details

SNP ID
rs142569261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5290987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTCACTGTGCATATTGTTCTC[A/G]TGCCTTTTCAGATCAGATGCCTTGG
Phenotype
MIM: 602126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB14 PubMed Links

Gene Details

Gene
ZBTB14
Gene Name
zinc finger and BTB domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143823.2 1383 Silent Mutation CAC,CAT H407H NP_001137295.1
NM_001243702.1 1383 Silent Mutation CAC,CAT H407H NP_001230631.1
NM_001243704.1 1383 Silent Mutation CAC,CAT H407H NP_001230633.1
NM_003409.4 1383 Silent Mutation CAC,CAT H407H NP_003400.2

View Full Product Details