Product Details

SNP ID
rs145703814
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:45985467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGTTTCTGAAAGAAAATAACA[A/C]AGAAATTTCCTCTGAATGAAAGGTG
Phenotype
MIM: 616046
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PSTPIP2 PubMed Links

Gene Details

Gene
PSTPIP2
Gene Name
proline-serine-threonine phosphatase interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024430.3 Intron NP_077748.3
XM_011526253.2 Intron XP_011524555.1
XM_011526254.1 Intron XP_011524556.1
XM_011526256.1 Intron XP_011524558.1

View Full Product Details