Product Details

SNP ID
rs137964332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47258567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTCCCGGTCTCAGGCAGCCAC[A/G]CTCGATTTGAAATCAAAGGAGGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC9 PubMed Links

Gene Details

Gene
CCDC9
Gene Name
coiled-coil domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015603.2 219 Silent Mutation ACA,ACG T4T NP_056418.1
XM_017026578.1 219 Silent Mutation ACA,ACG T4T XP_016882067.1

View Full Product Details