Product Details

SNP ID
rs138228528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12807220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCCAGACTCAAAGACCCTA[C/T]TGGAGAGCGAGCGGGAAAGGCTGTT
Phenotype
MIM: 600538 MIM: 606034
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRDX2 PubMed Links

Gene Details

Gene
PRDX2
Gene Name
peroxiredoxin 2
There are no transcripts associated with this gene.

Gene
RNASEH2A
Gene Name
ribonuclease H2 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006397.2 459 Silent Mutation CTG,TTG L72L NP_006388.2
XM_006722619.2 459 Silent Mutation CTG,TTG L28L XP_006722682.1

View Full Product Details