Product Details

SNP ID
rs138357652
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38326767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATGCGTTGGGGCTGGGACCCCC[C/G]GCGGGCGGCCTGCTGGCACTTGGTG
Phenotype
MIM: 613452 MIM: 603939
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CATSPERG PubMed Links

Gene Details

Gene
CATSPERG
Gene Name
cation channel sperm associated auxiliary subunit gamma
There are no transcripts associated with this gene.

Gene
KCNK6
Gene Name
potassium two pore domain channel subfamily K member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004823.1 1169 Missense Mutation CCG,CGG P166R NP_004814.1
XM_011527526.1 1169 Missense Mutation CCG,CGG P32R XP_011525828.1
XM_011527527.1 1169 Intron XP_011525829.1
XM_017027506.1 1169 Intron XP_016882995.1

View Full Product Details