Product Details

SNP ID
rs139073171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2098092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCATCTCCTGCAACAACTGCAC[A/G]GACTCGCACGTCGCCTGCTTTGGCT
Phenotype
MIM: 607246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3D1 PubMed Links

Gene Details

Gene
AP3D1
Gene Name
adaptor related protein complex 3 delta 1 subunit
There are no transcripts associated with this gene.

Gene
IZUMO4
Gene Name
IZUMO family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031735.2 536 Silent Mutation ACA,ACG T146T NP_001026905.2
NM_001039846.1 536 Silent Mutation ACA,ACG T146T NP_001034935.1
XM_005259480.3 536 Silent Mutation ACA,ACG T146T XP_005259537.2
Gene
MOB3A
Gene Name
MOB kinase activator 3A
There are no transcripts associated with this gene.

View Full Product Details