Product Details

SNP ID
rs140079108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47260394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCTCCCCGAAAGGGCCGCTCAG[A/T]GGAGAAGGAGAACGTGGCAGTGGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC9 PubMed Links

Gene Details

Gene
CCDC9
Gene Name
coiled-coil domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015603.2 389 Missense Mutation GAG,GTG E61V NP_056418.1
XM_017026578.1 389 Missense Mutation GAG,GTG E61V XP_016882067.1

View Full Product Details