Product Details

SNP ID
rs140312407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53140434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTAGGCTTGAACAGATACTAAA[C/G]GGTTTCCCACACTCATATGGTTTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF347 PubMed Links

Gene Details

Gene
ZNF347
Gene Name
zinc finger protein 347
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172674.1 2824 Silent Mutation CCC,CCG P799P NP_001166145.1
NM_001172675.1 2824 Silent Mutation CCC,CCG P799P NP_001166146.1
NM_032584.2 2824 Silent Mutation CCC,CCG P798P NP_115973.2
XM_005259335.4 2824 Silent Mutation CCC,CCG P798P XP_005259392.1
XM_005259336.4 2824 Silent Mutation CCC,CCG P740P XP_005259393.1
XM_017027384.1 2824 Silent Mutation CCC,CCG P798P XP_016882873.1
XM_017027385.1 2824 Silent Mutation CCC,CCG P740P XP_016882874.1
XM_017027386.1 2824 Silent Mutation CCC,CCG P740P XP_016882875.1
Gene
ZNF415
Gene Name
zinc finger protein 415
There are no transcripts associated with this gene.

View Full Product Details