Product Details

SNP ID
rs140839810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:307276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCGGATCCCCGCTGAGGTCCA[C/T]ATGGCTGGAGATGAGGGGCAGCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929060 PubMed Links

Gene Details

Gene
LOC101929060
Gene Name
uncharacterized LOC101929060
There are no transcripts associated with this gene.

Gene
MIER2
Gene Name
MIER family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005259582.4 2052 Missense Mutation ATG,GTG M489V XP_005259639.1
XM_006722769.3 2052 Missense Mutation ATG,GTG M482V XP_006722832.1
XM_011528072.2 2052 Missense Mutation ATG,GTG M489V XP_011526374.1
XM_011528073.1 2052 Missense Mutation ATG,GTG M487V XP_011526375.1
XM_011528074.2 2052 Missense Mutation ATG,GTG M482V XP_011526376.1
XM_011528075.2 2052 Missense Mutation ATG,GTG M451V XP_011526377.1
XM_011528077.2 2052 Missense Mutation ATG,GTG M451V XP_011526379.1
XM_011528081.2 2052 Missense Mutation ATG,GTG M411V XP_011526383.1
XM_011528082.2 2052 Missense Mutation ATG,GTG M403V XP_011526384.1

View Full Product Details