Product Details

SNP ID
rs140941620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48616098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACCGGAGAGCAGGACAGTGCC[A/G]CAGCCCTTAGGGGAGTCCAGGGCCA
Phenotype
MIM: 604179 MIM: 609932 MIM: 607092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83E PubMed Links

Gene Details

Gene
FAM83E
Gene Name
family with sequence similarity 83 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017708.3 677 Intron NP_060178.2
Gene
RPL18
Gene Name
ribosomal protein L18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000979.3 677 Silent Mutation TGC,TGT C134C NP_000970.1
NM_001270490.1 677 Silent Mutation TGC,TGT C105C NP_001257419.1
Gene
SPACA4
Gene Name
sperm acrosome associated 4
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
There are no transcripts associated with this gene.

View Full Product Details