Product Details

SNP ID
rs141575653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3751527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGGGCAGTGGACGATGCTGAG[C/T]GTCACCGACGTCTGCGACTTCGTCT
Phenotype
MIM: 604262 MIM: 612689
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APBA3 PubMed Links

Gene Details

Gene
APBA3
Gene Name
amyloid beta precursor protein binding family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004886.3 1621 Silent Mutation NP_004877.1
XM_006722950.3 1621 Silent Mutation XP_006723013.1
XM_006722951.3 1621 Silent Mutation XP_006723014.1
Gene
TJP3
Gene Name
tight junction protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267560.1 1621 Intron NP_001254489.1
NM_001267561.1 1621 Intron NP_001254490.1

View Full Product Details