Product Details
- SNP ID
-
rs142122989
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37564676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCACACTGGACACATGTATAGGG[C/T]TTCTCACCAGTATGGATCCTTTGAT
- Phenotype
-
MIM: 613903
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF540
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16973890,rs4802029] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF540
- Gene Name
- zinc finger protein 540
- Gene
- ZNF571
- Gene Name
- zinc finger protein 571
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290314.1 |
2519 |
Silent Mutation |
AAA,AAG |
K584K |
NP_001277243.1 |
NM_001321272.1 |
2519 |
Silent Mutation |
AAA,AAG |
K584K |
NP_001308201.1 |
NM_016536.4 |
2519 |
Silent Mutation |
AAA,AAG |
K584K |
NP_057620.3 |
XM_011527024.2 |
2519 |
Silent Mutation |
AAA,AAG |
K587K |
XP_011525326.1 |
XM_017026855.1 |
2519 |
Silent Mutation |
AAA,AAG |
K626K |
XP_016882344.1 |
XM_017026856.1 |
2519 |
Silent Mutation |
AAA,AAG |
K587K |
XP_016882345.1 |
XM_017026857.1 |
2519 |
Silent Mutation |
AAA,AAG |
K587K |
XP_016882346.1 |
XM_017026858.1 |
2519 |
Silent Mutation |
AAA,AAG |
K550K |
XP_016882347.1 |
XM_017026859.1 |
2519 |
Silent Mutation |
AAA,AAG |
K550K |
XP_016882348.1 |
XM_017026860.1 |
2519 |
Silent Mutation |
AAA,AAG |
K520K |
XP_016882349.1 |
- Gene
- ZNF571-AS1
- Gene Name
- ZNF571 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details