Product Details
- SNP ID
-
rs142205221
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4325456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAATGGGGGTCACGTAGTTCTCTT[C/G]CTGGTTCGGTAGTGGGGGCAGTGGG
- Phenotype
-
MIM: 609828
MIM: 607881
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FSD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1138253] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSD1
- Gene Name
- fibronectin type III and SPRY domain containing 1
There are no transcripts associated with this gene.
- Gene
- STAP2
- Gene Name
- signal transducing adaptor family member 2
View Full Product Details