Product Details

SNP ID
rs142842646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38411172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGAGCTCCGGGGGGTCCTGCAT[C/T]GCCCTTGGCCCCTGGCCTCTTCTTA
Phenotype
MIM: 607320
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
There are no transcripts associated with this gene.

Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146202.1 1907 Missense Mutation AAT,GAT N585D NP_001139674.1
NM_001146203.1 1907 Missense Mutation AAT,GAT N507D NP_001139675.1
NM_001146204.1 1907 Missense Mutation AAT,GAT N530D NP_001139676.1
NM_001146205.1 1907 Missense Mutation AAT,GAT N565D NP_001139677.1
NM_001146206.1 1907 Missense Mutation AAT,GAT N410D NP_001139678.1
NM_001146207.1 1907 Missense Mutation AAT,GAT N502D NP_001139679.1
NM_170604.2 1907 Missense Mutation AAT,GAT N599D NP_733749.1

View Full Product Details