Product Details

SNP ID
rs143766646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2717273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTGCTTCCCGGAGCGCTTGCC[A/G]TCGATGTTGAGGCTCATGTTCCGGC
Phenotype
MIM: 607862
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRAS1 PubMed Links

Gene Details

Gene
DIRAS1
Gene Name
DIRAS family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145173.3 857 Silent Mutation GAC,GAT D178D NP_660156.1
XM_011527713.2 857 Silent Mutation GAC,GAT D212D XP_011526015.1

View Full Product Details