Product Details

SNP ID
rs146029309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10875281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCTTGATTTGCAGCAGCGAAA[C/T]CCAAGTCCCTGCGTCTGTCTTCACT
Phenotype
MIM: 603934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf38 PubMed Links

Gene Details

Gene
C19orf38
Gene Name
chromosome 19 open reading frame 38
There are no transcripts associated with this gene.

Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199141.1 Intron NP_954592.1
XM_005259708.4 Intron XP_005259765.1
XM_011527638.2 Intron XP_011525940.1

View Full Product Details