Product Details

SNP ID
rs146318651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55014430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGCAAGACCCGTTCTGAGAGA[C/T]GAAAGGAGATTTGTTAGACCGCAGT
Phenotype
MIM: 605546
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GP6 PubMed Links

Gene Details

Gene
GP6
Gene Name
glycoprotein VI platelet
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083899.2 1539 Silent Mutation NP_001077368.2
NM_001256017.2 1539 UTR 3 NP_001242946.2
NM_016363.5 1539 UTR 3 NP_057447.5

View Full Product Details