Product Details

SNP ID
rs146345577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40759557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGACCCCGGCCACCAAGAAGGCT[A/G]TGCAAGGCGGGGGAGCCACCCCCGT
Phenotype
MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004596.4 928 Missense Mutation ATG,GTG M125V NP_004587.1

View Full Product Details