Product Details

SNP ID
rs146675788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44326759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCAAGTGCACACATGGGTACA[C/T]ATACACACACACAAAGCTAAACTTC
Phenotype
MIM: 603994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF112 PubMed Links

Gene Details

Gene
ZNF112
Gene Name
zinc finger protein 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083335.1 3529 UTR 3 NP_001076804.1
NM_013380.3 3529 UTR 3 NP_037512.3

View Full Product Details