Product Details

SNP ID
rs147700985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37565167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGGCCTTTCCACATTCCTTAC[A/G]TTTGTAGGGCTTCTCTCCGGTATGA
Phenotype
MIM: 613903
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF540 PubMed Links

Gene Details

Gene
ZNF540
Gene Name
zinc finger protein 540
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172225.2 2028 Intron NP_001165696.1
NM_001172226.2 2028 Intron NP_001165697.1
NM_152606.4 2028 Intron NP_689819.1
Gene
ZNF571
Gene Name
zinc finger protein 571
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290314.1 2028 Missense Mutation NP_001277243.1
NM_001321272.1 2028 Missense Mutation NP_001308201.1
NM_016536.4 2028 Missense Mutation NP_057620.3
XM_011527024.2 2028 Missense Mutation XP_011525326.1
XM_017026855.1 2028 Missense Mutation XP_016882344.1
XM_017026856.1 2028 Missense Mutation XP_016882345.1
XM_017026857.1 2028 Missense Mutation XP_016882346.1
XM_017026858.1 2028 Missense Mutation XP_016882347.1
XM_017026859.1 2028 Missense Mutation XP_016882348.1
XM_017026860.1 2028 Missense Mutation XP_016882349.1
Gene
ZNF571-AS1
Gene Name
ZNF571 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details