Product Details

SNP ID
rs149820790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17802568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGCCTGGAGGTCGAAGCCTCTC[A/G]GAGGTGACATTTCAGGGGAAGCCTA
Phenotype
MIM: 605863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GNT3 PubMed Links

Gene Details

Gene
B3GNT3
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014256.3 Intron NP_055071.2
XM_011527626.2 Intron XP_011525928.1

View Full Product Details